Fisherbrand™ Low-Retention Pipette Tips, Non-Filtered
Increase accuracy by reducing sample retention
Manufacturer:  Fisherbrand™ H10496RLRFIS
 View more versions of this product
Catalog No. 02-717-134
Specifications
Description
Tip, Pipet; Fisherbrand; Low Retention; Natural; 0.1-10uL; Racked, 10 x 96; Non-sterile, universal fit
  • Low retention surface minimizes samples loss
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Natural color
Description & Specifications
Specifications
0.1 to 10μL
Natural
Universal fit
Universal Fit
Nonsterile
Low retention
10 x 96/Rack