Please login to your online account to display your discounted pricing

Fisherbrand™ Low-Retention Pipette Tips, Nonfilter


Increase accuracy by reducing sample retention

Manufacturer: fisherbrand™  F113RNS15LRFIS

 View more versions of this product

Catalog No. 02-717-164

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

For Use With (Equipment) Universal Fit
Packaging 10 x 96/Rack
Sterility Sterile
Model Low retention
Color Natural
Volume (Metric) 150μL
Tip Style Beveled