Please login to your online account to display your discounted pricing

Fisherbrand™ Low-Retention Pipette Tips, Nonfilter


Increase accuracy by reducing sample retention

Manufacturer: fisherbrand™  111NXLR1SLRFIS

 View more versions of this product

Catalog No. 02-717-151

  • / Pack of 1000 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

For Use With (Equipment) Universal Fit
Sterility Sterile
Model Low Retention
Packaging Racked: 10 x 1000
Tip Style Extended Length
Volume (Metric) 1000μL
Color Natural