Please login to your online account to display your discounted pricing

Fisherbrand™ Low-Retention Pipette Tips, Nonfilter


Increase accuracy by reducing sample retention

Manufacturer: fisherbrand™  111NXLLRFIS

 View more versions of this product

Catalog No. 02-717-149

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Volume (Metric) 1000μL
Sterility Nonsterile
Color Natural
Model Low Retention
Tip Style Extended Length
Packaging Bulk Pack
For Use With (Equipment) Universal Fit