Fisherbrand™ Low-Retention Pipette Tips, Nonfilter
Increase accuracy by reducing sample retention
Description
Increase accuracy by reducing sample retention
- Low retention surface minimizes samples loss
- Certified by independent laboratory
- Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
- Natural color
Specifications
Low retentionSterile, Rack | |
Non-Filtered | |
Natural | |
Rack | |
Universal Fit |
200 μL | |
Sterile | |
Low retention | |
Beveled |