Fisherbrand™ Low-Retention Pipet Tips - Filtered
Increase accuracy by reducing sample retention
$163.93 - $186.28
Specifications
Sterility Sterile
Model Low retention
Color Natural
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Tip Style Packaging Volume (Metric) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Tip Style Packaging Volume (Metric) Price Quantity  
02-717-157 Fisherbrand™
F10496RS10LRFIS
Universal Fit Filter 10 x 96/Rack 10μL Pack of 960 Each for $163.93
Case of 5 Pack for $819.65
02-717-158 Fisherbrand™
FH102RS10LRFIS
Universal Fit Extended length, Filter 10 x 96/Rack 10μL Pack of 960 Each for $163.93
Case of 5 Pack for $819.65
02-717-161 Fisherbrand™
F113RNS20LRFIS
Universal Fit Beveled, Filter 10 x 96/Rack 20μL Pack of 960 Each for $163.93
Case of 5 Pack for $819.65
02-717-162 Fisherbrand™
F113RNS50LRFIS
Universal Fit Beveled, Filter 10 x 96/Rack 50μL Pack of 960 Each for $163.93
Case of 5 Pack for $819.65
02-717-165 Fisherbrand™
F10896RS2LRFIS
Universal Fit Beveled, Filter 10 x 96/Rack 200μL Pack of 960 Each for $163.93
Case of 5 Pack for $819.65
02-717-166 Fisherbrand™
F111R100S1LRFIS
Universal Fit Filter Racked: 10 x 100 1000μL Pack of 1000 Each for $186.28
Case of 5 Pack for $931.40
Specifications
Description
  • Low retention surface minimizes samples loss
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Natural color
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Sterile
Low retention
Natural