Please login to your online account to display your discounted pricing

Fisherbrand™ Low-Retention Pipet Tips - Filtered


Increase accuracy by reducing sample retention

Manufacturer: fisherbrand™  F10896RS2LRFIS

 View more versions of this product

Catalog No. 02-717-165

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Packaging 10 x 96/Rack
For Use With (Equipment) Universal Fit
Sterility Sterile
Model Low retention
Tip Style Beveled, Filter
Volume (Metric) 200μL
Color Natural