Please login to your online account to display your discounted pricing

Fisherbrand™ Low-Retention Pipet Tips - Filtered


Increase accuracy by reducing sample retention

Manufacturer: fisherbrand™  F113RNS50LRFIS

 View more versions of this product

Catalog No. 02-717-162

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Sterility Sterile
Packaging 10 x 96/Rack
For Use With (Equipment) Universal Fit
Model Low retention
Volume (Metric) 50μL
Tip Style Beveled, Filter
Color Natural