Please login to your online account to display your discounted pricing
Fisherbrand™ Low-Form Polypropylene Beakers
Smooth, tapered design facilitates easy handling and pouring
$50.89 - $198.93
Specifications
Autoclavable Yes
Material PP (Polypropylene)
Product Type Beaker
For Use With (Application) Easy handling and pouring
Graduated Yes
View More Specs
Products
Catalog Number Mfr. No. Graduations Capacity (Metric) Price Quantity    
02-591-27
  Accessories
fisherbrand™
5226550100F
20mL 100mL Pack of 10 Each for $97.39
02-591-25
fisherbrand™
5226550025F
5mL 25mL Pack of 10 Each for $77.96
02-591-26
  Accessories
fisherbrand™
5226550050F
10mL 50mL Pack of 10 Each for $77.96
02-591-29
  Accessories
fisherbrand™
5226550400F
50mL 400mL Pack of 10 Each for $179.89
02-591-30
  Accessories
fisherbrand™
5226550500F
50mL 500mL Pack of 10 Each for $198.93
02-591-33
  Accessories
fisherbrand™
5226552000F
200mL 2000mL Pack for $50.89
02-591-34
  Accessories
fisherbrand™
5226553000F
200mL 3000mL Pack for $58.61
02-591-35
  Accessories
fisherbrand™
5226555000F
500mL 5000mL Pack for $82.36
02-591-28
  Accessories
fisherbrand™
5226550250F
50mL 250mL Pack of 10 Each for $123.29
02-591-32
  Accessories
fisherbrand™
5226551000F
100mL 1000mL Pack of 5 Each for $141.79
02-591-31
  Accessories
fisherbrand™
5226550600F
50mL 600mL Pack of 5 Each for $111.39
Description & Specifications
Specifications
Autoclavable Yes
Material PP (Polypropylene)
Product Type Beaker
For Use With (Application) Easy handling and pouring
Graduated Yes
Includes Anti-lock
Scale Blue graduated scale
Temperature (English) Max. Operating 250°F
Temperature (Metric) Max. Operating 121°C
  • Resistant to most acids, bases, and many commonly used solvents
  • Shatter resistant
  • Easy-to-read, blue printed graduations
  • “No Flame” symbol and SPI recycling code Anti-lock feature prevents beakers from sticking together when stacked
  • Autoclavable
  • Withstand temperatures up to 121°C (250°F)