Fisherbrand™ Low-Form Dewar Flasks
Slightly curved bottom increases coolant capacity
$915.57 - $2,024.80
Specifications
Base Material Aluminum
For Use With (Application) Liquefied gases
Style Low-form cylindrical
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Height (English) Height (Metric) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Height (English) Height (Metric) Capacity (Metric) Price Quantity  
10-196-6 Accessories Fisher Scientific
POS8325
4.6 in. 11.7cm 0.35L Each for $915.57
10-196-7 Accessories Fisher Scientific
POS8335
4.33 in. 11cm 0.85L Each for $1,112.77
10-196-8 Accessories Fisher Scientific
POS8345
7.28 in. 18.5cm 2.5L Each for $2,024.80
Specifications
Description
  • Cylindrical style
  • For use with liquified gases
  • Vacuum tip—off tubulation is offset to minimize space between the bottom and magnetic stirrer
  • Protective aluminum base extends to full height of the Dewar
Description & Specifications
Specifications
Aluminum
Liquefied gases
Low-form cylindrical