Fisherbrand™ Low-Cost Cast-Iron Tripod-Base Supports
Finished with chemical-resistant, baked, black enamel
Manufacturer:  Fisherbrand™ 00407Q**
 View more versions of this product
Catalog No. 14-675BQ
Description
Description
Finished with chemical-resistant, baked, black enamel
  • Base is heavy, rugged cast iron for stability
  • Support rods and base precisely threaded to eliminate instability
Specifications
Specifications
Tripod Base Support
0.37 in.
Baked Black Enamel
20 in.
Each
Cast Iron
0.95cm
Support rods and base precisely threaded to eliminate instability
51cm
SDS
Documents
Accessories