Fisherbrand™ LoSUDS Liquid Glassware Detergent DFS Item
1 gallon jug detergent for cleaning glassware. Cuts grease and oils leaving glassware spotless.
Manufacturer:  Fisherbrand™ DET150
Catalog No. 13-641-730
Description
Description
 Bar Maid LoSUDS detergent can be used with all manual and electric washers to leave glassware crystal clear.
  • Cuts grease and oils
  • Leaves glassware spotless
  • Use only one ounce for every three gallons of water
  • Easy on hands
Specifications
Specifications
 Detergent for cleaning glassware
 1
 128fl. oz.
 All manual and electric glass washers
1 gal.
Jug
 Portable glass washers
 1EA