Fisherbrand™ Long-Stem Analytical Funnels
Molded polypropylene with external ridges and outside ribs to prevent vapor lock
Manufacturer:  Fisherbrand™ N3006000045
 View more versions of this product
Catalog No. 10-500-12
Specifications Description & Specifications
Specifications
Yes
0.77 oz., 23mL
4.8cm
8.3cm
5cm
0.77 oz.
Polypropylene
1.88 in. (48mm)
7cm
1.88 in.
3.26 in.
1.96 in.
23mL
58° Bowl