Please login to your online account to display your discounted pricing
Fisherbrand™ Locking-Lid Microcentrifuge Tubes with Polypropylene Snap-Cap™
Convenient, pierceable locking lids snap into place with one-handed operation
Manufacturer: fisherbrand™  L510GRDVFIS
 View more versions of this product
Catalog No. 02-681-288
  • / Pack of 500 Each
  • / Case of 10 Pack
Description & Specifications
Specifications
For Use With (Application) Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
Packaging Bag
Autoclavable Yes
Color Violet
Closure Type Snap
Closure Material Polypropylene
Max. Force 26000xG
Capacity (Metric) 1.5mL
Graduations 0.1mL
Material Polypropylene
Certifications/Compliance RNase, DNase-free
Accessories
Accessories for Catalog Number: 02681288
Item Mfr. No. Item Description Price Per Unit Quantity  
05-407-22
Fisherbrand™ Microcentrifuge Tube Accessory Micro Tube Tool
fisherbrand™
BIP4000
For use in capping or decapping Fisherbrand Microcentrifuge Tubes with Snap Caps Pack of 4 Each for $14.20
Case of 25 Pack for $355.00