Fisherbrand™ Locking-Lid Microcentrifuge Tubes with Polypropylene Snap-Cap™
Convenient, pierceable locking lids snap into place with one-handed operation
Manufacturer:  Fisherbrand™ L508GRDYFIS
 View more versions of this product
Catalog No. 02-681-295
Specifications Description & Specifications
Specifications
RNase, DNase-free
Snap
2mL
Polypropylene
Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
Bag
Polypropylene
Yes
Yellow
25000xG
0.1mL
Accessories