Please login to your online account to display your discounted pricing
Fisherbrand™ Locking-Lid Microcentrifuge Tubes with Polypropylene Snap-Cap™
Convenient, pierceable locking lids snap into place with one-handed operation
Manufacturer: fisherbrand™  L508GRDRFIS
 View more versions of this product
Catalog No. 02-681-292
  • / Pack of 500 Each
  • / Case of 10 Pack
Description & Specifications
Specifications
Closure Material Polypropylene
Color Red
Certifications/Compliance RNase, DNase-free
Closure Type Snap
Packaging Bag
Material Polypropylene
Max. Force 25000xG
Graduations 0.1mL
Capacity (Metric) 2mL
Autoclavable Yes
For Use With (Application) Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
Accessories
Accessories for Catalog Number: 02681292
Item Mfr. No. Item Description Price Per Unit Quantity  
05-407-22
Fisherbrand™ Microcentrifuge Tube Accessory Micro Tube Tool
fisherbrand™
BIP4000
For use in capping or decapping Fisherbrand Microcentrifuge Tubes with Snap Caps Pack of 4 Each for $14.20
Case of 25 Pack for $355.00