Fisherbrand™ Locking-Lid Microcentrifuge Tubes with Polypropylene Snap-Cap™
Convenient, pierceable locking lids snap into place with one-handed operation
Manufacturer:  Fisherbrand™ L502GRDG
 View more versions of this product
Catalog No. 02-681-276
Description
Description
Convenient, pierceable locking lids snap into place with one-handed operation
  • Polypropylene tubes with flat tops
  • Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
  • With graduation marks for volume estimation
  • RNase and DNase free
Centrifuges that develop excessive heat may damage this tube.
Specifications
Specifications
RNase, DNase-free
Snap
0.6mL
Polypropylene
Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
Bag
Polypropylene
Yes
Green
30000xG
0.1mL
Documents
Accessories