Fisherbrand™ Locking Dialysis Membrane Clamps
Provides a leakproof seal.
$120.30 - $133.23
Specifications
Quantity 10 Pack
For Use With (Application) Do not float, stay locked until released
Material Polyamide 6.6 Dupont (Nylon)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
21-152-13 Fisherbrand™
CB1070
Dialysis Membrane Clamp, Up to 65mm sealing width Pack of 10 Each for $133.23
21-153-100 Fisherbrand™
2115212
Dialysis Membrane Clamp, Up to 46mm sealing width Pack of 10 Each for $120.30
Specifications
Description
  • Stay locked until released
  • Do not float
  • Constructed of Polyamide 6.6 from DuPont (NYLON)
  • Available in 2 sizes: 46mm or 65mm
  • 10 clamps/pkg
Description & Specifications
Specifications
10 Pack
Do not float, stay locked until released
Polyamide 6.6 Dupont (Nylon)