Please login to your online account to display your discounted pricing
Fisherbrand™ Liquid Proof Scale Papers, 0.004 in. Thick
Works for mixing then weighing on the same sheet
Manufacturer: fisherbrand™  20605705
 View more versions of this product
Catalog No. 12-587-105
  • / Pack of 200 Each
Description & Specifications
Specifications
Dimensions (L x W) 25.4 x 50.8mm (1 x 2 in.)
Quantity 200/Pk.
Product Type Liquid proof scale paper