Fisherbrand™ Lens Paper
For cleaning glass lenses
Manufacturer:  Fisherbrand™ 20205117
 View more versions of this product
Catalog No. 11-995
Description
Description
For cleaning glass lenses
Booklet
  • Never scratches glass lenses, objectives, or other delicate surfaces
  • Doesn't become harsh; doesn't readily collect dust or grease
  • Virtually lint-free sheets are glued and bound in a booklet for easy removal
Dispenser Box
  • Same paper as above, but packaged in a convenient dispenser box for fast, single-sheet removal individual sheets measure 5L × 7 in.W (13 × 18cm)
Specifications
Specifications
Book of 6 x 8 in. sheets
100 sheets/book, 12 books/pack; 1200
Lens cleaning paper
For cleaning Glass lenses