Please login to your online account to display your discounted pricing

Fisherbrand™ Leather Driver's Gloves

Provide abrasion resistance and excellent dexterity

$100.21 - $100.21

Specifications

Certifications ISO9001:2008
Color Natural
Sterile Yes
For Use With (Application) construction, machine operation, light duty welding, utility work, farming, roping and many other general applications
Glove Cuff Style Slip-on
View More Specs
Products
Catalog Number Mfr. No. Hem Color Thumb Style Material Size Price Quantity    

19-042128

 
protective industrial
68118S
Red Straight Premium grade cowhide leather, cotton Small CALL CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Certifications ISO9001:2008
Color Natural
Sterile Yes
For Use With (Application) construction, machine operation, light duty welding, utility work, farming, roping and many other general applications
Glove Cuff Style Slip-on
Powdered Yes

  • Ideal for construction, machine operation, utility work, farming, roping, and other general applications
  • Top-grain leather is available with straight thumb or keystone thumb to provide additional wear and greater comfort
  • Launderable