Please login to your online account to display your discounted pricing
Fisherbrand™ Latex Gloves
Fisherbrand™ Latex Gloves offer excellent dexterity and superior tactile sensitivity.
Manufacturer: fisherbrand™  113945DD
 View more versions of this product
Catalog No. 11-394-5DD
  • / Pack of 100 Each
  • / Case of 10 Pack
Description & Specifications
Specifications
Material Latex
Powdered No
Size X-Large
  • Excellent for health care and laboratory applications due to exceptional dexterity and tactile sensitivity of this medical grade glove
  • Provides water-tight barrier protection against infectious and hazardous materials
  • Extra-strong rolled cuff makes donning easier and prevents roll down
  • Ambidextrous and available in a wide range of sizes for superior fit and comfort
Accessories
Accessories for Catalog Number: 113945DD
Item Mfr. No. Item Description Price Per Unit Quantity