Please login to your online account to display your discounted pricing
Fisherbrand™ Latex Gloves
Fisherbrand™ Latex Gloves offer excellent dexterity and superior tactile sensitivity.
Manufacturer: fisherbrand™  11394125B
 View more versions of this product
Catalog No. 11-394-125B
  • / Pack of 100 Each
  • / Case of 10 Pack
Description & Specifications
Specifications
Material Latex
Powdered Lightly-powdered
Size Medium
  • Excellent for health care and laboratory applications due to exceptional dexterity and tactile sensitivity of this medical grade glove
  • Provides water-tight barrier protection against infectious and hazardous materials
  • Extra-strong rolled cuff makes donning easier and prevents roll down
  • Ambidextrous and available in a wide range of sizes for superior fit and comfort
Accessories
Accessories for Catalog Number: 11394125B
Item Mfr. No. Item Description Price Per Unit Quantity  
04-355-21
Decon™ Proguard™ Professional Hand Cream
decon laboratories
DC9516
Specially formulated lotion moisturizes dry, cracked hands and guards against latex glove irritation Each for $71.42
Case of 6 Each for $428.52
11-127-47A
S-Curve™ Acrylic Glove Dispensers
s-curve™
EBD1000
Wall-mountable, clear acrylic Each for $38.60
Case of 10 Each for $353.40