Please login to your online account to display your discounted pricing
Fisherbrand™ Latex-free Nitrile Gloves
Fisherbrand™ Latex-free Nitrile Gloves Offer superior strenghth and puncture resisistance.
$11.39 - $12.52
Specifications
Material Nitrile
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
27-058-50
fisherbrand™
2705850
X-Small Pack of 100 Each for $12.52
Case of 10 Pack for $119.80
27-058-51
fisherbrand™
2705851
Small Pack of 100 Each for $11.86
Case of 10 Pack for $113.40
27-058-52
fisherbrand™
2705852
Medium Pack of 100 Each for $11.86
Case of 10 Pack for $113.40
27-058-53
fisherbrand™
2705853
Large Pack of 100 Each for $11.86
Case of 10 Pack for $113.40
27-058-54
fisherbrand™
2705854
X-Large Pack of 100 Each for $11.39
Case of 10 Pack for $108.90
Description & Specifications
Specifications
Material Nitrile
  • Contains no latex proteins that can cause allergic reactions 4mil thickness for greater barrier protection and resists permeation by a wider range of chemicals
  • Superior tensile strength, puncture resistance and durability
  • Fully textured surface and fingertips provide superior grip in both wet and dry conditions
  • Beaded cuff for added strength and easier donning
  • Precise glove fit, comfortable feel, increased flexibility and freedom of movement