Please login to your online account to display your discounted pricing
Fisherbrand™ Large-Orifice Pipet Tips
Manufacturer: fisherbrand™  H11896RSFIS
 View more versions of this product
Catalog No. 02-707-134
  • / Pack of 960 Each
  • / Case of 5 Pack
Description & Specifications
Specifications
Packaging Hinged Rack, 10 x 96
Length (English) 2 in.
Length (Metric) 5cm
For Use With (Equipment) Biohit Proline*, Proline Electronic ; BrandTech* Transferpette* ; Costar* Onepette* ; Eppendorf* 3130 ; Eppendorf Reference 2000 ; Eppendorf Research 2100 ; Eppendorf Single Channel ; Finnpipette* Digital ; Finnpipette, Labsystems, Single-Channel Fixed ;
Sterility Sterile
Tip Style Large Orifice
Volume (Metric) 1 to 200μL
Color Yellow