Fisherbrand™ Fisherbrand™ Large-Orifice Pipet Tips
Fisherbrand™ Large-Orifice Pipet Tips helps you pipet difficult liquids.
Description
Genomic/Wide Orifice assists in pipetting small volumes of viscous materials.
- Assist in pipetting small volumes of viscous materials
- Help eliminate shearing of genomic DNA and fragile cells
Specifications
100 to 1000 μL | |
Natural | |
6.6cm | |
Genomic/Wide Orifice | |
Costar Onepette, Eppendorf, Eppendorf Reference 2000, Eppendorf Research 2100, Eppendorf Research 3130, Eppendorf Reference 2000, Eppendorf Research 2100, Eppendorf, Thermo Scientific Finnpipette Digital |
Non-sterile | |
2.625 in. | |
Rack | |
LR, 10 x 100 |