Fisherbrand™ Large-Orifice Pipet Tips, 1 to 200μL
Manufacturer:  Fisherbrand™ 119NXLFIS
 View more versions of this product
Catalog No. 02-707-143
Description
Description
  • Assist in pipetting small volumes of viscous materials
  • Help eliminate shearing of genomic DNA and fragile cells
Specifications
Specifications
Sartorius™ Biohit™ Proline™, Sartorius™ Biohit™ Proline™ Electronic, Transferpette™, Costar™ Onepette™, Eppendorf™ Research™ 3130, Eppendorf™ Reference™ 2000, Eppendorf™ Research™ 2100, Eppendorf™, Thermo Scientific™ Finnpipette™ Digital
Genomic/Wide Orifice
Non-sterile
3.4 in.
Sartorius™ Biohit™ Proline™, Sartorius™ Biohit™ Proline™ Electronic, Transferpette™, Costar™ Onepette™, Eppendorf™ Research™ 3130, Eppendorf™ Reference™ 2000, Eppendorf™ Research™ 2100, Eppendorf™ Pipettor, Thermo Scientific™ Finnpipette™ Digital
Bulk
100 to 1000μL
Natural
8.6cm
Documents