Fisherbrand™ Labware Washer Brushes
Heavy-duty, long-life brushes are designed to give optimum cleaning results
Manufacturer:  Fisherbrand™ BRS975
 View more versions of this product
Catalog No. 13-641-710
Description
Description
  • Some Brushes are stiffer on top and softer on the sides for better cleaning
  • Variety of shapes and sizes to fit most labware
  • Made in the U.S.A.
Specifications
Specifications
21.6cm
Standard Tall Brush
Labware washer designed to give optimum cleaning results
8.5 in.
Standard tall - 8.5 in.
Documents