missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Laboratory Carts and Tables

Fisherbrand Laboratory Carts offer versatile choices for holding or transporting a variety of lab supplies, equipment and containers.

Manufacturer:  Fisherbrand™ LCN8019002

 View more versions of this product

Catalog No. 15-252-400



Description

Description

Fisherbrand Laboratory Carts offer versatile choices for holding or transporting a variety of lab supplies, equipment and containers.

  • 4 in. diameter casters, 2 with toe locks
  • Ready-to-roll, full factory assembly
  • Ergonomic, extruded rubber handle grips
  • Durable Fisherbrand blue, powder-coated 18 gauge welded tubular steel frame
  • ID tag
  • 5 year warranty
  • Supports loads up to 400 lbs.

Stainless Steel Cart features two stainless steel pains that are 2 in. deep on three sides with one open side for easy transfer of equipment from countertop to cart.

2-Basket Glassware Cart, used to carry glassware and small instruments in two vinyl-coated wire baskets, includes removable plastoc drip pan to catch spills. Each basket measures 32 x 18 x 4 in. h.

4-Basket Glassware Cart, used to carry glassware and small instruments in four vinyl-coated wire baskets, includes removable plastic drip pan to catch spills. Each basket measures 16 x 18 x 4 in. h.

Pan Cart, used for transport of wet or soiled labware, features 2 in. deep powder-coated steel, removable watertight pans lined with gray rubber mats to prevent shifting.

Portable Table features two open shelves that provide easy transfer of equipment from countertop to cart. Chemical Cart features two open shelves with rigid PVC extrusions on two sides to prevent containers from sliding.

Warranty

5-years

Specifications

Specifications

36.38 in.
19 in.
400 lb.
Blue
34.88 in.
48.3cm
2
Two removable polished type 304 Stainless Steel pans that are 2 in. deep on three sides with one open side
26.3kg
Laboratory cart
Stainless Steel
181kg
92.4cm
88.6cm
34.88 x 19 x 36.38 in. (88.6 x 48.3 x 92.4cm)
Versatile for holding or transporting variety of lab supplies, equipment and containers
58 lb.
Documents
Promotions

Promotions