Fisherbrand™ Lab Jacks
Precise height adjustment with the turn of a knob.
Manufacturer:  Fisherbrand™ 19090005
 View more versions of this product
Catalog No. 14-673-53
Description
Description
Precise height adjustment with the turn of a knob.
  • For supporting hotplates, baths, glassware and other items requiring stable regulation
  • 14-673-51, 14-673-52, 14-673-53 feature threaded holes to accommodate an 18.87 in. (48cm) long aluminum rod for mounting ring stands and other lab apparatus
Order Info
Rod available separately; contact your Fisher Customer Service Representative.
Warranty
Two years
Specifications
Specifications
3.5 to 20 in.
12 in.
Lab Jack
12 in.
30cm
Supporting hotplates, baths, glassware
8.9 to 50.8cm
30cm
12 in.
30cm
143 lb.
Stainless Steel, Anodized Aluminum Top
SDS
Documents
Accessories