Fisherbrand™ Lab Jacks
Precise height adjustment with the turn of a knob.
Manufacturer:  Fisherbrand™ 19090004
 View more versions of this product
Catalog No. 14-673-52
Description
Description
Precise height adjustment with the turn of a knob.
  • For supporting hotplates, baths, glassware and other items requiring stable regulation
  • 14-673-51, 14-673-52, 14-673-53 feature threaded holes to accommodate an 18.87 in. (48cm) long aluminum rod for mounting ring stands and other lab apparatus
Order Info
Rod available separately; contact your Fisher Customer Service Representative.
Warranty
Two years
Specifications
Specifications
2.25 to 11 in.
8 in.
Lab Jack
8 in.
20cm
Supporting hotplates, baths, glassware
5.7 to 28cm
20cm
8 in.
20cm
79 lb.
Stainless Steel, Anodized Aluminum Top
SDS
Documents
Accessories