missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Hazard Warning Label Refills

Printed in safety colors.

Manufacturer:  Fisherbrand™ 15932

 View more versions of this product

Catalog No. 15-932




Description

Description

For use with Fisherbrand™ Lab Hazard Warning Label Dispenser (cat. no. 15931).

  • Tape rolls preprinted with warnings in safety colors: POISON (yellow on black); CORROSIVE (black on yellow); FLAMMABLE (red on white); and RADIOACTIVE (red on yellow)
  • Serrated tear-off edge for easy tape separation
  • Approximately 200 imprints per roll

Order Info

Each tape roll measures 0.50 in. W x 22 yd. L (13mm x 20m)

Specifications

Specifications

Assorted, Assorted
Printed in safety colors
20.3m
Tape
4 Case
Poison
57 yd.
0.5 in.
Poison Warning Refill Tape
Documents