Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  A22821FIS

 View more versions of this product

Catalog No. 02-707-216

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Robotic Tip
Volume (Metric) 250μL
Sterility Nonsterile
Color Blue Tint
Autoclavable Yes
Tip Style Automation, Disposable, Racked
Material Polypropylene
Packaging Rack
Quantity 96 Tips/Rack
For Use With (Application) Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures
For Use With (Equipment) Beckman™ Coulter™ Multimek™ 96 robotic lab automation workstation