Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  14296RSFIS

 View more versions of this product

Catalog No. 02-707-240

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Material Polypropylene
Product Type Robotic Tip
Quantity 96 Tips/Rack
For Use With (Equipment) Beckman™ Coulter™ Biomek™ 1000 and 2000 robotic lab automation workstation
Sterility Sterile
Autoclavable Yes
Tip Style Automation, Disposable, Racked
Packaging Rack
For Use With (Application) Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures
Volume (Metric) 5 to 250μL
Color Natural