Fisherbrand™ Lab Automation Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures
Manufacturer:  Fisherbrand™ 14296RFIS
 View more versions of this product
Catalog No. 02-707-239
Specifications Description & Specifications
Specifications
Robotic Tip
Nonsterile
Yes
Polypropylene
96 Tips/Rack
Beckman™ Coulter™ Biomek™ 1000 and 2000 robotic lab automation workstation
5 to 250μL
Natural
Automation, Disposable, Racked
Rack
Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures