Fisherbrand™ Lab Automation Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures
Description
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures
Carbon-black, liquid-level-sensing tips are required by many automated systems. They are constructed of a highly conductive material that is necessary to detect presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.
Disposable tips for robotic workstations — provide maximum performance, accuracy, and reproducibility in laboratory automation procedures. Available in clear, blue tint, or carbon-black-conductive polypropylene.
Features
- Ideal for demanding HTS procedures
- Provide maximum performance, accuracy, and reproducibility in lab automation procedures
- Engineered to ensure proper fit and function with several standard and customized automated liquid-handling systems
- Choose tips in an assortment of volume sizes and styles
Specifications
50 μL | |
Natural | |
Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures | |
Automation | |
BioTek Precision, Tecan TeMo, Tecan GenMate |
Sterile | |
Autoclavable | |
Rack | |
Single-use | |
Polypropylene |