Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  16396RFIS

 View more versions of this product

Catalog No. 02-707-344

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Quantity 96 Tips/Rack
For Use With (Equipment) BioTek™ Precision™ and Tecan™ TEMO and Tecan GenMate™ robotic lab automation workstation
Material Polypropylene
Sterility Nonsterile
Product Type Robotic Tip
Packaging Rack
Autoclavable Yes
Tip Style Automation, Disposable, Racked
Volume (Metric) 50μL
Color Natural
For Use With (Application) Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures