Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  16296RSFIS

 View more versions of this product

Catalog No. 02-707-343

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Sterility Sterile
For Use With (Equipment) BioTek™ Precision™ and Tecan™ TEMO and Tecan GenMate™ robotic lab automation workstation
Product Type Robotic Tip
Material Polypropylene
Quantity 96 Tips/Rack
For Use With (Application) Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures
Color Natural
Packaging Rack
Tip Style Automation, Disposable, Racked
Autoclavable Yes
Volume (Metric) 200μL