Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  16196RSFIS

 View more versions of this product

Catalog No. 02-707-341

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Robotic Tip
Volume (Metric) 100μL
Sterility Sterile
Color Natural
Autoclavable Yes
Tip Style Automation, Disposable, Racked
Material Polypropylene
Packaging Rack
Quantity 96 Tips/Rack
For Use With (Application) Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures
For Use With (Equipment) BioTek™ Precision™ and Tecan™ TEMO and Tecan GenMate™ robotic lab automation workstation