Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  B14196RSFIS

 View more versions of this product

Catalog No. 02-707-274

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Material Polypropylene
Quantity 96 Tips/Rack
Product Type Robotic Tip
Sterility Sterile
For Use With (Equipment) Packard™ MultiProbe™ I and II robotic lab automation workstation
Autoclavable Yes
For Use With (Application) Liquid-level-sensing to detect presence or absence of sufficient liquid, provide maximum performance, accuracy and reproducibility in laboratory automation procedures
Volume (Metric) 20μL
Packaging Rack
Color Black