Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  14196RFIS

 View more versions of this product

Catalog No. 02-707-236

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

For Use With (Equipment) Beckman™ Coulter™ Biomek™ 1000 and 2000 robotic lab automation workstation
Material Polypropylene
Quantity 96 Tips/Rack
Product Type Robotic Tip
Sterility Nonsterile
For Use With (Application) Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures
Volume (Metric) 1 to 20μL
Autoclavable Yes
Color Natural
Tip Style Automation, Disposable, Racked
Packaging Rack