Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Nonfilter Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  15996RFIS

 View more versions of this product

Catalog No. 02-707-224

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Robotic Tip
Material Polypropylene
Quantity 96 Tips/Rack
For Use With (Equipment) Zymark™ RapidPlate™ 96 and 96/384 robotic lab automation workstation
Sterility Nonsterile
Autoclavable Yes
Tip Style Automation, Disposable, Nonfilter, Racked
Packaging Rack
For Use With (Application) Provide maximum performance, accuracy, and reproducibility in laboratory automation procedures
Volume (Metric) 100μL
Color Natural