Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Nonfilter Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  17596RSFIS

 View more versions of this product

Catalog No. 02-707-260

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Quantity 96 Tips/Rack
Sterility Sterile
Material Polypropylene
Product Type Robotic Tip
For Use With (Equipment) Tecan™ Genesis™ robotic lab automation workstation
Tip Style Automation, Disposable, Nonfilter, Racked
Autoclavable Yes
Packaging Rack
Color Natural
Volume (Metric) 1000μL
For Use With (Application) Provide maximum performance, accuracy, and reproducibility in laboratory automation procedures