Fisherbrand™ Lab Automation Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures.
Description
Tips are constructed of a highly conductive material necessary to detect the presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.
- Disposable tips for robotic workstations provide maximum performance, accuracy and reproducibility in laboratory automation procedures
- Available in clear, blue tint, or carbon-black-conductive polypropylene
- Carbon-black, liquid-level-sensing tips are required by many automated systems
Specifications
200 μL | |
Sterile | |
Autoclavable | |
Rack | |
Single-use | |
Polypropylene |
Non-Filtered | |
Natural | |
Provide maximum performance, accuracy, and reproducibility in laboratory automation procedures | |
Automation | |
Tecan Genesis |