Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Nonfilter Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  17196RFIS

 View more versions of this product

Catalog No. 02-707-244

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Robotic Tip
Volume (Metric) 20μL
Sterility Nonsterile
Color Natural
Autoclavable Yes
Tip Style Automation, Disposable, Nonfilter, Racked
Material Polypropylene
Packaging Rack
Quantity 96 Tips/Rack
For Use With (Application) Provide maximum performance, accuracy, and reproducibility in laboratory automation procedures
For Use With (Equipment) Tecan™ Genesis™ robotic lab automation workstation