Fisherbrand™ Lab Automation Filter Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures
Manufacturer:  Fisherbrand™ F142R9S180FIS
 View more versions of this product
Catalog No. 02-707-242
Description
Description
Tips are constructed of a highly conductive material necessary to detect the presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.
  • Disposable tips for robotic workstations provide maximum performance, accuracy and reproducibility in laboratory automation procedures
  • Available in clear, blue tint, or carbon-black-conductive polypropylene
  • Carbon-black, liquid-level-sensing tips are required by many automated systems
Specifications
Specifications
Beckman Coulter™ Biomek™ 1000/2000
Automation
Filtered
Natural
PP (Polypropylene)
Beckman Coulter™ Biomek™ 1000/2000
Rack
180μL
Sterile
Yes
Demanding HTS procedures, lab automation procedures; fit several standard and customized automated liquid-handling systems
Documents