Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Filter Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  F141R9S20FIS

 View more versions of this product

Catalog No. 02-707-241

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Sterility Sterile
For Use With (Equipment) Beckman™ Biomek™ 1000/2000
Product Type Filtered Automation Tip
Material Polypropylene
Packaging Rack
Volume (Metric) 20μL
Autoclavable Yes
Color Natural
For Use With (Application) Demanding HTS procedures, lab automation procedures; fit several standard and customized automated liquid-handling systems