Fisherbrand™ Lab Automation Filter Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures
Manufacturer:  Fisherbrand™ F173R9S200FIS
 View more versions of this product
Catalog No. 02-707-265
Description
Description
Tips are constructed of a highly conductive material necessary to detect the presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.
  • Disposable tips for robotic workstations provide maximum performance, accuracy and reproducibility in laboratory automation procedures
  • Available in clear, blue tint, or carbon-black-conductive polypropylene
  • Carbon-black, liquid-level-sensing tips are required by many automated systems
Specifications
Specifications
Tecan™ Genesis™
Automation
Filtered
Natural
PP (Polypropylene)
Tecan™ Genesis™
Rack
200μL
Sterile
Yes
Demanding HTS procedures, lab automation procedures; fit several standard and customized automated liquid-handling systems
Documents