Please login to your online account to display your discounted pricing

Fisherbrand™ Lab Automation Filter Pipet Tips


Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Manufacturer: fisherbrand™  F160R9S150FIS

 View more versions of this product

Catalog No. 02-707-228

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Sterility Nonsterile
For Use With (Equipment) Zymark™ RapidPlate™ 96 and 96/384
Material Polypropylene
Product Type Filtered Automation Tip
Color Natural
For Use With (Application) Demanding HTS procedures, lab automation procedures; fit several standard and customized automated liquid-handling systems
Volume (Metric) 150μL
Autoclavable Yes
Packaging Rack