Fisherbrand™ Kolmer Serological Pipets
For Kolmer Complement Fixation technique.
Manufacturer:  Fisherbrand™ 13676M
 View more versions of this product
Catalog No. 13-676M
Description
Description
  • Accuracy printed on each pipet
  • Borosilicate glass with permanent amber markings
  • Tuf-Temp™ treated to reduce chipping
  • All Fisherbrand Reusable Serological Pipets also available as PersonaPet™ personalized pipets
Compliance
Calibrated “To Deliver” to baseline.
Specifications
Specifications
±0.06mL
Orange
Yes
Serological Pipet
Borosilicate Glass
To deliver to baseline
Orange
0.01mL
10mL